missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphine, 95%, ACROS Orgaincs™

Brand:  Acros Organics 320970250

Additional Details : CAS Number : 829-85-6 Weight : 0.07500kg Transport : UN number : 2845 Chem class : 4.2 Pack group : I


Quantity 25 g

 View more versions of this product

Product Code. 10023993

  • 1599.00 DKK / 25g

Estimated Shipment: 02-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

829-85-6
C12H11P
186.19

Specifications

Diphenylphosphine
110°C
1.6240 to 1.6260
280.0°C
829-85-6
C12H11P
15, 3364
186.19
1.0700 g/mL
Glass bottle
1.07
25 g
95%
C6H5PHC6H5
Solubility aq. soln.: insoluble.
186.19
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diphenylphosphine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Pyrophoric liquid Category 1

Hazard Statement
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H250-Catches fire spontaneously if exposed to air.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates