missing translation for 'onlineSavingsMsg'
Få mere at vide

Diphenylamine hydrochloride, 99%, BLD Pharm™

Artikelnummer. 31004672
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
25 g
100 g
Pakningsstørrelse:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
31004672 100 g 100g
30992321 25 g 25g
30989388 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31004672 Leverandør BLD Pharm Leverandørnr. BD125861100g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale Diphenylamine hydrochloride
CAS 537-67-7
Molekylær formel C12H12ClN
MDL nummer MFCD00042007
Mængde 100 g
SMIL [H]Cl.C1(NC2=CC=CC=C2)=CC=CC=C1
Molekylvægt (g/mol) 205.68
Procent renhed 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.