missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Diphenyl(piperidin-4-yl)methanol (Azacyclonol; Pipradrol 4-Isomer) 1.0 mg/ml in Methanol, LoGiCal
Beskrivelse
Diphenyl(piperidin-4-yl)methanol (Azacyclonol; Pipradrol 4-Isomer) 1.0 mg/ml in Methanol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pheniramine Maleate |
| CAS | 132-20-7 |
| Analyt- eller komponentnavne | Pheniramine Maleate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H20 N2 . C4 H4 O4 |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Brompheniramine Maleate Imp. C (EP), Brompheniramine Imp. C (EP), Dexchlorpheniramine Maleate Imp. A (EP), Dexchlorpheniramine Imp. A (EP), (3 RS)-N,N-Dimethyl-3-phenyl-3-(pyridin-2-yl)propan-1-amine Maleate, N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1), N,N-Dimethyl-gamma-phenyl-2-pyridinepropanamine (2 Z)-2-butenedioate (1:1) |
| InChI formel | InChI=1 S/C16H20N2.C4H4O4/c1-18(2)13-11-15(14-8-4-3-5-9-14)16-10-6-7-12-17-16;5-3(6)1-2-4(7)8/h3-10,12,15 H,11,13H2,1-2H3;1-2 H,(H,5,6)(H,7,8)/b;2-1- |
| SMIL | CN(C)CCC(c1ccccc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC navn | (Z)-but-2-enedioic acid;N,N-dimethyl-3-phenyl-3-pyridin-2-ylpropan-1-amine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?