Få mere at vide
Diphenyl Phosphate-d10, TRC
Artikelnummer. 30371963
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30371963 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371963 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4920021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diphenyl Phosphate-d10
CAS 1477494-97-5
Renhedsgrad noter HPLC
Molekylær formel C12 D10 H O4 P
Mængde 1 mg
Synonym Phosphoric Acid Diphenyl Ester-d10, Diphenyl Hydrogen Phosphate-d10, NSC 6518-d10, Phenyl Phosphate-d10
InChI formel InChI=1S/C12H11O4P/c13-17(14,15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H,(H,13,14)/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(OP(=O)(O)Oc2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H]
IUPAC navn bis(2,3,4,5,6-pentadeuteriophenyl) hydrogen phosphate
Molekylvægt (g/mol) 260.25
Formel vægt 260.1023
Procent renhed >95