missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl carbonate, 99%, Thermo Scientific Chemicals

Catalog Number p-7082092
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
102-09-0
C13H10O3
214.22
MFCD00003037
ROORDVPLFPIABK-UHFFFAOYSA-N
carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate
7597
CHEBI:34722
diphenyl carbonate
C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
102-09-0
C13H10O3
214.22
MFCD00003037
ROORDVPLFPIABK-UHFFFAOYSA-N
carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate
7597
CHEBI:34722
diphenyl carbonate
C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2

Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.

Solubility
Soluble in ethanol, diethyl ether, carbon tetrachloride, acetic acid. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 76°C to 81°C
Density 1.3
Boiling Point 301°C to 302°C
Flash Point 168°C (334°F)
Odor Mild
Quantity 250 g
Beilstein 1074863
Merck Index 14,3602
Solubility Information Soluble in ethanol,diethyl ether,carbon tetrachloride,acetic acid. Insoluble in water.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material Diphenyl carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'