missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Diphenoxylate Hydrochloride, Mikromol
Description
Diphenoxylate Hydrochloride, Mikromol
Specifications
Specifications
| Kemisk navn eller materiale | Diphenoxylate hydrochloride |
| CAS | 3810-80-8 |
| Analyt- eller komponentnavne | Diphenoxylate hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C30 H32 N2 O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Diphenoxylate hydrochloride, Ethyl 1-(3-cyano-3,3-diphenylpropyl)-4-phenylpiperidine-4-carboxylate hydrochloride, 4-Piperidinecarboxylic acid, 1-(3-cyano-3,3-diphenylpropyl)-4-phenyl-, ethyl ester, hydrochloride (1:1), 4-Piperidinecarboxylic acid, 1-(3-cyano-3,3-diphenylpropyl)-4-phenyl-, ethyl ester, monohydrochloride (9CI), Isonipecotic acid, 1-(3-cyano-3,3-diphenylpropyl)-4-phenyl-, ethyl ester, hydrochloride (6CI,7CI), Isonipecotic acid, 1-(3-cyano-3,3-diphenylpropyl)-4-phenyl-, ethyl ester, monohydrochloride (8CI), 1-(3-Cyano-3,3-diphenylpropyl)-4-phenylpiperidine-4-carboxylic acid ethyl ester hydrochloride, 2,2-Diphenyl-4-(4-carbethoxy-4-phenyl-1-piperidino)butyronitrile hydrochloride, 2,2-Diphenyl-4-(4-carboxy-4-phenyl-1-piperidino)butyronitrile ethyl ester hydrochloride, R 1132, R 1132 (antiperistaltic) |
| InChI formel | InChI=1S/C30H32N2O2.ClH/c1-2-34-28(33)29(25-12-6-3-7-13-25)18-21-32(22-19-29)23-20-30(24-31,26-14-8-4-9-15-26)27-16-10-5-11-17-27;/h3-17H,2,18-23H2,1H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCOC(=O)C1(CCN(CCC(C#N)(c2ccccc2)c3ccccc3)CC1)c4ccccc4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?