Få mere at vide
Diphenoxylate Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Diphenoxylate hydrochloride |
| CAS | 3810-80-8 |
| Analyt- eller komponentnavne | Diphenoxylate hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C30 H32 N2 O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Diphenoxylate hydrochloride, Ethyl 1-(3-cyano-3,3-diphenylpropyl)-4-phenylpiperidine-4-carboxylate hydrochloride, 4-Piperidinecarboxylic acid, 1-(3-cyano-3,3-diphenylpropyl)-4-phenyl-, ethyl ester, hydrochloride (1:1), 4-Piperidinecarboxylic acid, 1-(3-cyano-3,3-diphenylpropyl)-4-phenyl-, ethyl ester, monohydrochloride (9CI), Isonipecotic acid, 1-(3-cyano-3,3-diphenylpropyl)-4-phenyl-, ethyl ester, hydrochloride (6CI,7CI), Isonipecotic acid, 1-(3-cyano-3,3-diphenylpropyl)-4-phenyl-, ethyl ester, monohydrochloride (8CI), 1-(3-Cyano-3,3-diphenylpropyl)-4-phenylpiperidine-4-carboxylic acid ethyl ester hydrochloride, 2,2-Diphenyl-4-(4-carbethoxy-4-phenyl-1-piperidino)butyronitrile hydrochloride, 2,2-Diphenyl-4-(4-carboxy-4-phenyl-1-piperidino)butyronitrile ethyl ester hydrochloride, R 1132, R 1132 (antiperistaltic) |
| InChI formel | InChI=1S/C30H32N2O2.ClH/c1-2-34-28(33)29(25-12-6-3-7-13-25)18-21-32(22-19-29)23-20-30(24-31,26-14-8-4-9-15-26)27-16-10-5-11-17-27;/h3-17H,2,18-23H2,1H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCOC(=O)C1(CCN(CCC(C#N)(c2ccccc2)c3ccccc3)CC1)c4ccccc4 |
| Vis mere |