missing translation for 'onlineSavingsMsg'
Learn More

Diphenhydramine, TRC

Product Code. 30373628
Change view
Click to view available options
Mængde:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30373628 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373628 Supplier Toronto Research Chemicals Supplier No. TRCD4871282.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Diphenhydramine
CAS 58-73-1
Renhedsgrad noter HPLC
Molekylær formel C17H21NO
Mængde 2.5 g
Synonym Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl- (7CI,8CI), 2-(Diphenylmethoxy)-N,N-dimethylethanamine, 2-(Benzhydryloxy)-N,N-dimethylethylamine, 2-(Diphenylmethoxy)-N,N-dimethylethylamine, Benzhydramine, DPH, Dimedrol base, Diphenhydramine, FAR 90X2, N,N-Dimethyl-2-(diphenylmethoxy)ethylamine, N-[2-(Diphenylmethoxy)ethyl]-N,N-dimethylamine, NSC 665800, O-Benzhydryldimethylaminoethanol, Probedryl, α-(2-Dimethylaminoethoxy)diphenylmethane, β-Dimethylaminoethanol diphenylmethyl ether, β-Dimethylaminoethylbenzhydrylether
InChI formel InChI=1S/C17H21NO/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,17H,13-14H2,1-2H3
SMIL CN(C)CCOC(c1ccccc1)c2ccccc2
IUPAC navn 2-benzhydryloxy-N,N-dimethylethanamine
Molekylvægt (g/mol) 255.361
Formel vægt 255.162
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.