Få mere at vide
Diphenhydramine-d6 Hydrochloride, TRC
Artikelnummer. 30377544
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30377544 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377544 Leverandør Toronto Research Chemicals Leverandørnr. TRCD48690250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diphenhydramine-d6 Hydrochloride
CAS 1189986-72-8
Renhedsgrad noter HPLC
Molekylær formel C17H16D6ClNO
Mængde 50 mg
Synonym 2-(Diphenylmethoxy)-N,N-dimethylethanamine-d6 Hydrochloride, 2-(Benzhydryloxy)-N,N-dimethylethylamine-d6 Hydrochloride, Allergina-d6, Benadryl-d6, Dibondrin-d6, Noctomin-d6
InChI formel InChI=1S/C17H21NO.ClH/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12,17H,13-14H2,1-2H3;1H/i1D3,2D3;
SMIL Cl.[2H]C([2H])([2H])N(CCOC(c1ccccc1)c2ccccc2)C([2H])([2H])[2H]
IUPAC navn 2-benzhydryloxy-N,N-bis(trideuteriomethyl)ethanamine;hydrochloride
Molekylvægt (g/mol) 297.85
Formel vægt 297.1767
Procent renhed >95