Få mere at vide
Dioxypyramidon, Mikromol™
Artikelnummer. 30461867
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30461867 4 x 25 mg
30051701 100 mg
30052141 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30461867 Leverandør Mikromol Leverandørnr. MM1049.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Dioxypyramidon, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dioxypyramidon
CAS 519-65-3
Analyt- eller komponentnavne Dioxypyramidon
Forsendelsestilstand Room Temperature
Molekylær formel C13 H17 N3 O3
Certificeringer/compliance ISO 17025
Synonym Acetic acid, 2-(dimethylamino)-2-oxo-, 2-acetyl-2-methyl-1-phenylhydrazide, Acetic acid, (dimethylamino)oxo-, 2-acetyl-2-methyl-1-phenylhydrazide (9CI), Semioxamazide, 1-acetyl-1,5,5-trimethyl-2-phenyl- (6CI,7CI,8CI), 1-Acetyl-1,5,5-trimethyl-2-phenylsemioxamazide, Dioxoaminopyrine, Dioxyaminopyrine, Dioxypyramidon, alpha-(Dimethylamidooxalyl)-beta,beta-methylacetylphenylhydrazine
InChI formel InChI=1S/C13H17N3O3/c1-10(17)15(4)16(11-8-6-5-7-9-11)13(19)12(18)14(2)3/h5-9H,1-4H3
Løsningstype Neat
SMIL CN(C)C(=O)C(=O)N(N(C)C(=O)C)c1ccccc1
IUPAC navn 2-(N-[acetyl(methyl)amino]anilino)-N,N-dimethyl-2-oxoacetamide
Molekylvægt (g/mol) 263.29
Mængde 4 x 25 mg
Formel vægt 263.127 g/mol