missing translation for 'onlineSavingsMsg'
Få mere at vide

Dioxindolyl-L-alanine (Mixture of Diastereomers), TRC

Artikelnummer. 30005620
Change view
Click to view available options
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30005620 50 mg
30006820 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005620 Supplier Toronto Research Chemicals Supplier No. TRCD48685550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184955-21-3
Molekylær formel C11 H12 N2 O4
Molekylvægt (g/mol) 236.22
Synonym (αS)-α-Amino-2,3-dihydro-3-hydroxy-2-oxo-1H-indole-3-propanoic Acid, [3(S)]-α-Amino-2,3-dihydro-3-hydroxy-2-oxo-1H-indole-3-propanoic Acid;
IUPAC navn (2S)-2-amino-3-(3-hydroxy-2-oxo-1H-indol-3-yl)propanoic acid
SMIL N[C@@H](CC1(O)C(=O)Nc2ccccc12)C(=O)O

Specifications

Kemisk navn eller materiale (2S)-2-amino-3-(3-hydroxy-2-oxo-1H-indol-3-yl)propanoic acid
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C11H12N2O4/c12-7(9(14)15)5-11(17)6-3-1-2-4-8(6)13-10(11)16/h1-4,7,17H,5,12H2,(H,13,16)(H,14,15)/t7-,11?/m0/s1
Formel vægt 236.0797
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.