Få mere at vide
Diosmin EP Impurity F, Mikromol™
Artikelnummer. 30051815
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051815 25 mg
30051905 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051815 Leverandør Mikromol Leverandørnr. MM0816.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diosmin EP Impurity F
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diosmetin
CAS 520-34-3
Analyt- eller komponentnavne Diosmetin
Forsendelsestilstand Room Temperature
Molekylær formel C16 H12 O6
Certificeringer/compliance ISO 9001
Synonym 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one (Diosmetin), 4H-1-Benzopyran-4-one, 5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-, Diosmetin (6CI), Flavone, 3',5,7-trihydroxy-4'-methoxy- (7CI,8CI), 3',5,7-Trihydroxy-4'-methoxyflavone, 4'-Methylluteolin, 5,7,3'-Trihydroxy-4'-methoxyflavone, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, Diosmetine, Diosmetol, Luteolin 4'-methyl ether, Pillon, Disomin Imp. F (EP), Diosmin Impurity F
InChI formel InChI=1S/C16H12O6/c1-21-13-3-2-8(4-10(13)18)14-7-12(20)16-11(19)5-9(17)6-15(16)22-14/h2-7,17-19H,1H3
SMIL COc1ccc(cc1O)C2=CC(=O)c3c(O)cc(O)cc3O2
IUPAC navn 5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chromen-4-one
Molekylvægt (g/mol) 300.26
Mængde 25 mg
Formel vægt 300.0634 g/mol