Få mere at vide
Dioscin, TRC
Artikelnummer. 30133324
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133324 50 mg
30133069 25 mg
30006887 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133324 Leverandør Toronto Research Chemicals Leverandørnr. TRCD48523050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19057-60-4
Molekylær formel C45 H72 O16
Molekylvægt (g/mol) 869.06
Synonym (+)-Dioscin, 3-O-β-D-α-L-Rhamnopyranosyl(1→4)-[α-L-rhamnopyranosyl(1→2)]-β-D-glucopyranoside-diosgenin, Collettiside III, Dioscine, Diosgenin 3-O-α-L-Rhamnopyranosyl-(1→2)-[α-L-rhamnopyranosyl-(1→4)]-β-D-glucopyranoside, Diosgenin 3-O-α-L-Rhamnopyranosyl-(1→4)-[α-L-rhamnopyranosyl-(1→2)]-β-D-glucopyranoside, Diosgenin bis-α-L-rhamnopyranosyl-(1→2 and 1→4)-β-D-glucopyranoside, Paris III, Polyphyllin III, (3β,25R)-Spirost-5-en-3-yl O-6-deoxy-α-L-mannopyranosyl-(1→2)-O-[6-deoxy-α-L-mannopyranosyl-(1→4)]-β-D-glucopyranoside
IUPAC navn (2S,2'S,3R,3'R,4R,4'R,5R,5'R,6S,6'S)-6,6'-(((2R,3S,4S,5R,6R)-4-hydroxy-2-(hydroxymethyl)-6-(((4S,5'R,6aR,6bS,8aS,8bR,9S,10R,11aS,12aS,12bS)-5',6a,8a,9-tetramethyl-1,3,3',4,4',5,5',6,6a,6b,6',7,8,8a,8b,9,11a,12,12a,12b-icosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-yl)oxy)tetrahydro-2H-pyran-3,5-diyl)bis(oxy))bis(2-methyltetrahydro-2H-pyran-3,4,5-triol)
SMIL O[C@H]1[C@H](O[C@H]2[C@H](O)[C@@H](O[C@H](O[C@H]3C)[C@@H]([C@H](O)[C@H]3O)O)[C@H](O[C@@H]4CC5=CC[C@@]6([H])[C@@](C7)([H])[C@@](CC[C@]6([H])[C@@]5(C)CC4)(C)[C@]8([H])[C@@]7([H])O[C@@](CC[C@H]9C)(OC9)[C@H]8C)O[C@@H]2CO)O[C@@H](C)[C@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale Dioscin
Mængde 50 mg
InChI formel InChI=1S/C45H72O16/c1-19-9-14-45(54-18-19)20(2)30-28(61-45)16-27-25-8-7-23-15-24(10-12-43(23,5)26(25)11-13-44(27,30)6)57-42-39(60-41-36(52)34(50)32(48)22(4)56-41)37(53)38(29(17-46)58-42)59-40-35(51)33(49)31(47)21(3)55-40/h7,19-22,24-42,46-53H,8-18H2,1-6H3/t19-,20+,21+,22+,24+,25-,26+,27+,28+,29-,30+,31+,32+,33-,34-,35-,36-,37+,38-,39-,40+,41+,42-,43+,44+,45-/m1/s1
Formel vægt 868.482