Få mere at vide
Dioctyltinmercaptide (~90%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Plastic additive 23 |
| CAS | 15571-58-1 |
| Molekylær formel | C36 H72 O4 S2 Sn |
| Mængde | 10 mg |
| Synonym | 10-Ethyl-4,4-dioctyl-7-oxo-8-oxa-3,5-dithia-4-stannatetradecanoic acid 2-ethylhexyl ester, [(Dioctylstannylene)dithio]diacetic acid bis(2-ethylhexyl) ester, Bis[(carboxymethyl)thio]dioctyltin bis(2-ethylhexyl) ester, 17MOK, Advastab 17MOK, Advastab 17MOL, Bis(2-ethylhexyl thioglycolato)dioctyltin, Bis(2-ethylhexyl) [(dioctylstannylene)dithio]diacetate, CAT 214, Dioctyltin bis(2-ethylhexyl thioglycolate), KS 2000A10, Reatinor 847, Thermolite 895, Plastic additive 23 (EP) |
| InChI formel | InChI=1S/2C10H20O2S.2C8H17.Sn/c2*1-3-5-6-9(4-2)7-12-10(11)8-13;2*1-3-5-7-8-6-4-2;/h2*9,13H,3-8H2,1-2H3;2*1,3-8H2,2H3;/q;;;;+2/p-2 |
| SMIL | CCCCCCCC[Sn](CCCCCCCC)(SCC(=O)OCC(CC)CCCC)SCC(=O)OCC(CC)CCCC |
| IUPAC navn | 2-ethylhexyl 2-[[2-(2-ethylhexoxy)-2-oxoethyl]sulfanyl-dioctylstannyl]sulfanylacetate |
| Molekylvægt (g/mol) | 751.79 |
| Formel vægt | 752.3894 |