missing translation for 'onlineSavingsMsg'
Få mere at vide

Dioctyltinmercaptide (>85%), TRC

Artikelnummer. 30373310
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30373310 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Artikelnummer. 30373310 Leverandør Toronto Research Chemicals Leverandørnr. TRCD48215050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Plastic additive 23
CAS 15571-58-1
Molekylær formel C36 H72 O4 S2 Sn
Mængde 50 mg
Synonym 10-Ethyl-4,4-dioctyl-7-oxo-8-oxa-3,5-dithia-4-stannatetradecanoic acid 2-ethylhexyl ester, [(Dioctylstannylene)dithio]diacetic acid bis(2-ethylhexyl) ester, Bis[(carboxymethyl)thio]dioctyltin bis(2-ethylhexyl) ester, 17MOK, Advastab 17MOK, Advastab 17MOL, Bis(2-ethylhexyl thioglycolato)dioctyltin, Bis(2-ethylhexyl) [(dioctylstannylene)dithio]diacetate, CAT 214, Dioctyltin bis(2-ethylhexyl thioglycolate), KS 2000A10, Reatinor 847, Thermolite 895, Plastic additive 23 (EP)
InChI formel InChI=1S/2C10H20O2S.2C8H17.Sn/c2*1-3-5-6-9(4-2)7-12-10(11)8-13;2*1-3-5-7-8-6-4-2;/h2*9,13H,3-8H2,1-2H3;2*1,3-8H2,2H3;/q;;;;+2/p-2
SMIL CCCCCCCC[Sn](CCCCCCCC)(SCC(=O)OCC(CC)CCCC)SCC(=O)OCC(CC)CCCC
IUPAC navn 2-ethylhexyl 2-[[2-(2-ethylhexoxy)-2-oxoethyl]sulfanyl-dioctylstannyl]sulfanylacetate
Molekylvægt (g/mol) 751.79
Formel vægt 752.3894
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.