Få mere at vide
Dioctylsulfosuccinic Acid Sodium Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dioctylsulfosuccinic Acid Sodium Salt |
| CAS | 1639-66-3 |
| Molekylær formel | C20 H37 O7 S . Na |
| Mængde | 2.5 g |
| Synonym | Dioctylsulfosuccinic Acid Sodium Salt, 2-Sulfobutanedioic Acid 1,4-Dioctyl Ester Sodium Salt, Sulfobutanedioic Acid 1,4-Dioctyl Ester Sodium Salt, Sulfosuccinic Acid 1,4-Dioctyl Ester Sodium Salt, Sulfosuccinic Acid Dioctyl Ester Sodium Salt, Octyl Alcohol Sulfosuccinate (1:2) Sodium Salt, Dioctyl Sodiosulfosuccinate, MKC, NSC 7779, Neocol SW 30, Penetrating T, Sodium 1,2-Bis(octyloxycarbonyl)-1-ethanesulfonate, Sodium Di-n-octyl Sulfosuccinate, Surfacpol 203, Texapon DOS, Sodium 1,4-Bis(octyloxy)-1,4-dioxobutane-2-sulfonate |
| InChI formel | InChI=1S/C20H38O7S.Na/c1-3-5-7-9-11-13-15-26-19(21)17-18(28(23,24)25)20(22)27-16-14-12-10-8-6-4-2;/h18H,3-17H2,1-2H3,(H,23,24,25);/q;+1/p-1 |
| SMIL | [Na+].CCCCCCCCOC(=O)CC(C(=O)OCCCCCCCC)S(=O)(=O)[O-] |
| IUPAC navn | sodium;1,4-dioctoxy-1,4-dioxobutane-2-sulfonate |
| Molekylvægt (g/mol) | 444.558 |
| Formel vægt | 444.216 |