Få mere at vide
Dinotefuran DN Hydrochloride, TRC
Artikelnummer. 30133067
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133067 100 mg
30136147 50 mg
30012010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133067 Leverandør Toronto Research Chemicals Leverandørnr. TRCD484610100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 939773-22-5
Molekylær formel C7H15N3O . HCl
Molekylvægt (g/mol) 193.67
Synonym N-Methyl-N'-[(tetrahydro-3-furanyl)methyl]guanidine Hydrochloride;1-Methyl-3-((tetrahydrofuran-3-yl)methyl)guanidine Hydrochloride;
SMIL Cl.CNC(=N)NCC1CCOC1
Tekniske data
Kemisk navn eller materiale Dinotefuran DN Hydrochloride
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C7H15N3O.ClH/c1-9-7(8)10-4-6-2-3-11-5-6;/h6H,2-5H2,1H3,(H3,8,9,10);1H
Formel vægt 193.0982
Procent renhed >95%