Få mere at vide
Dimetridazole-d3, TRC
Artikelnummer. 30378533
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30378533 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378533 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4796625MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dimetridazole D3
CAS 64678-69-9
Molekylær formel C5 2H3 H4 N3 O2
Mængde 5 mg
Synonym 1H-Imidazole, 2-methyl-1-(methyl-d3)-5-nitro- (9CI), Dimetridazole-d3, Dimetridazole D3 (N-methyl D3)
InChI formel InChI=1S/C5H7N3O2/c1-4-6-3-5(7(4)2)8(9)10/h3H,1-2H3/i2D3
SMIL [2H]C([2H])([2H])n1c(C)ncc1[N+](=O)[O-]
IUPAC navn 2-methyl-5-nitro-1-(trideuteriomethyl)imidazole
Molekylvægt (g/mol) 144.15
Formel vægt 144.0727