Få mere at vide
Dimetridazole-d3 (1-methyl-d3), CDN
Artikelnummer. 30311672
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311672 0.05 g
30321192 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311672 Leverandør CDN Leverandørnr. CDND79030.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dimetridazole D3
CAS 64678-69-9
Molekylær formel C5 2H3 H4 N3 O2
Mængde 0.05 g
Synonym 1 H-Imidazole, 2-methyl-1-(methyl-d3)-5-nitro- (9 CI), Dimetridazole-d3, Dimetridazole D3 (N-methyl D3)
InChI formel InChI=1 S/C5H7N3O2/c1-4-6-3-5(7(4)2)8(9)10/h3H,1-2H3/i2D3
SMIL [2 H]C([2 H])([2 H])n1c(C)ncc1[N+](=O)[O-]
IUPAC navn 2-methyl-5-nitro-1-(trideuteriomethyl)imidazole
Molekylvægt (g/mol) 144.15
Formel vægt 144.0727 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity