Få mere at vide
Dimetindene Maleate, LoGiCal
Artikelnummer. 30462694
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462694 10 mg
1 options
Artikelnummer. 30462694 Leverandør LoGiCal Leverandørnr. LGCFOR0595.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dimetindene maleate
CAS 3614-69-5
Analyt- eller komponentnavne Dimetindene maleate
Forsendelsestilstand Room Temperature
Molekylær formel C20 H24 N2 . C4 H4 O4
Certificeringer/compliance ISO 9001
Synonym Dimetindene maleate, N,N-Dimethyl-2-[3-[(RS)-1-(pyridin-2-yl)ethyl]-1H-inden-2-yl]ethanamine (Z)-butenedioate, 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, (2Z)-2-butenedioate (1:1), 1H-Indene-2-ethanamine, N,N-dimethyl-3-[1-(2-pyridinyl)ethyl]-, (Z)-2-butenedioate (1:1), Pyridine, 2-[1-[2-(dimethylaminoethyl)inden-3-yl]ethyl]-, maleate (6CI), Pyridine, 2-[1-[2-[2-(dimethylamino)ethyl]inden-3-yl]ethyl]-, maleate (1:1) (7CI,8CI), 2-[1-[2-(2-Dimethylaminoethyl)inden-3-yl]ethyl]pyridine maleate, Dimethinden maleate, Dimethindene maleate, Dimethpyrindene maleate, Fenistil, Fenistil-retard, Fenostil, Forhistal maleate, NSC 107677, Su-6518
InChI formel InChI=1S/C20H24N2.C4H4O4/c1-15(19-10-6-7-12-21-19)20-17(11-13-22(2)3)14-16-8-4-5-9-18(16)20;5-3(6)1-2-4(7)8/h4-10,12,15H,11,13-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Løsningstype Neat
SMIL CC(C1=C(CCN(C)C)Cc2ccccc12)c3ccccn3.OC(=O)\C=C/C(=O)O
IUPAC navn (Z)-but-2-enedioic acid;N,N-dimethyl-2-[3-(1-pyridin-2-ylethyl)-1H-inden-2-yl]ethanamine
Industry Type Clinical Testing
Molekylvægt (g/mol) 408.49
Mængde 10 mg
Formel vægt 408.2049 g/mol