Learn More
Dimethylphenylphosphine, 96%, Thermo Scientific Chemicals
Catalog Number p-7023933
Click to view available options
Quantity:
1 g
5 g
672-66-2
C8H11P
138.15
MFCD00008509
HASCQPSFPAKVEK-UHFFFAOYSA-N
dimethylphenylphosphine, dimethyl phenyl phosphine, dimethyl phenyl phosphane, me2pph, phenyldimethylphosphine, pme2ph, phosphine, dimethylphenyl, pubchem6420, dimethyl phenylphosphine
69597
CHEBI:30671
dimethyl(phenyl)phosphane
CP(C)C1=CC=CC=C1
672-66-2
C8H11P
138.15
MFCD00008509
HASCQPSFPAKVEK-UHFFFAOYSA-N
dimethylphenylphosphine, dimethyl phenyl phosphine, dimethyl phenyl phosphane, me2pph, phenyldimethylphosphine, pme2ph, phosphine, dimethylphenyl, pubchem6420, dimethyl phenylphosphine
69597
CHEBI:30671
dimethyl(phenyl)phosphane
CP(C)C1=CC=CC=C1
Dimethylphenylphosphine is used as a ligand in transition metal complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethylphenylphosphine is used as a ligand in transition metal complexes.
Solubility
Soluble in organic solvents.
Notes
Air sensitive. Store away from oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY
Density 0.971
Boiling Point 75°C to 79°C (12 mmHg)
Flash Point 50°C (122°F)
Odor Unpleasant
Refractive Index 1.563
Quantity 1 g
UN Number UN1993
Beilstein 742064
Sensitivity Air Sensitive
Solubility Information Soluble in organic solvents.
Formula Weight 138.15
Percent Purity 97%
Chemical Name or Material Dimethylphenylphosphine