Få mere at vide
Dimethyldimyristylammonium Bromide, TRC
Artikelnummer. 30388518
Skift visning
Mængde:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30388518 500 mg
30376895 250 mg
30385739 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30388518 Leverandør Toronto Research Chemicals Leverandørnr. TRCD481283500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68105-02-2
Molekylær formel C30H64BrN
Molekylvægt (g/mol) 518.74
Synonym 1-Tetradecanaminium, N,N-dimethyl-N-tetradecyl-, bromide (9CI), Di-n-tetradecyldimethylammonium bromide, Dimethyldimyristylammonium bromide, Dimethylditetradecylammonium bromide, Dimyristyldimethylammonium bromide, Ditetradecyldimethylammonium bromide, Dimethylditetradecylammonium Bromide, N,N-dimethyl-N-tetradecyl-1-Tetradecanaminium bromide, Di-n-tetradecyldimethylammonium bromide, Dimethylditetradecylammonium bromide, Dimyristyldimethylammonium bromide, Ditetradecyldimethylammonium bromide
IUPAC navn dimethyl-di(tetradecyl)azanium;bromide
SMIL [Br-].CCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCC
Tekniske data
Kemisk navn eller materiale Dimethyldimyristylammonium Bromide
Mængde 500 mg
InChI formel InChI=1S/C30H64N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31(3,4)30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-30H2,1-4H3;1H/q+1;/p-1
Formel vægt 517.4222