Få mere at vide
Dimethylammonium Dimethylcarbamate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dimethylammonium Dimethylcarbamate |
| CAS | 4137-10-4 |
| Molekylær formel | C3H7NO2 . x C2H7N |
| Mængde | 5 mL |
| Synonym | Dimethylcarbamic Acid, compd. with Dimethylamine, Dimethylcarbamic Acid, , compd. with Dimethylamine, N-Methylmethanamine Dimethylcarbamate, Dimcarb, Dimethylamine, ,N-dimethylcarbamate, Dimethylamine, compd. with Dimethylcarbamic Acid |
| InChI formel | InChI=1S/C3H7NO2.C2H7N/c1-4(2)3(5)6;1-3-2/h1-2H3,(H,5,6);3H,1-2H3 |
| SMIL | CNC.CN(C)C(=O)O |
| IUPAC navn | dimethylcarbamic acid;N-methylmethanamine |
| Molekylvægt (g/mol) | 134.18 |
| Formel vægt | 134.1055 |