Få mere at vide
Dimethylamine-d3 Hydrochloride, TRC
Artikelnummer. 30371444
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30371444 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371444 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4467322.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dimethylamine methyl-d3 hydrochloride
CAS 120033-84-3
Molekylær formel C2 2H3 H4 N . Cl H
Mængde 2.5 mg
Synonym Methan-d3-amine, N-methyl-, hydrochloride (9CI), N-Methylmethan-d3-amine hydrochloride, Dimethyl-1,1,1-d3-amine HCl, Dimethylamine-d3 Hydrochloride, Dimethylamine-D3 (N-methyl-D3) Hydrochloride
InChI formel InChI=1S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])NC
IUPAC navn 1,1,1-trideuterio-N-methylmethanamine;hydrochloride
Molekylvægt (g/mol) 84.56
Formel vægt 84.0534