Learn More
Dimethyl (trityloxyimino)malonate, 98%, Thermo Scientific™
Catalog Number 15408198
Click to view available options
Quantity:
250 mg
1 g
5 g
82874-83-7
(Trityloxyimino)malonic acid dimethyl ester; Dimethyl 2-[(triphenylmethoxy)imino]malonate
82874-83-7
(Trityloxyimino)malonic acid dimethyl ester; Dimethyl 2-[(triphenylmethoxy)imino]malonate
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 403.43
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Dimethyl (trityloxyimino)malonate