missing translation for 'onlineSavingsMsg'
Få mere at vide

Dimethyl Succinate-d4, TRC

Artikelnummer. 30375393
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30375393 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375393 Leverandør Toronto Research Chemicals Leverandørnr. TRCD472816100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Dimethyl Succinate-d4
CAS 30994-23-1
Molekylær formel C6 D4 H6 O4
Mængde 100 mg
Synonym Butanedioic-2,2,3,3-d4 acid, 1,4-dimethyl ester (ACI), 1,4-Dimethyl butanedioate-2,2,3,3-d4 (ACI), Butanedioic-d4 acid, dimethyl ester (9CI), Succinic-d4 acid, dimethyl ester (7CI, 8CI), Butanedioic-2,2,3,3-d4 acid 1,4-dimethyl ester, Succinic-d4 acid dimethyl ester, Dimethyl Succinate-d4, Succinic acid dimethyl ester-D4 (succinic-D4)
InChI formel InChI=1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3/i3D2,4D2
SMIL [2H]C([2H])(C(=O)OC)C([2H])([2H])C(=O)OC
IUPAC navn dimethyl 2,2,3,3-tetradeuteriobutanedioate
Molekylvægt (g/mol) 150.1659
Formel vægt 150.083
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.