missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Dimethyl phthalate-3,4,5,6-d4, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Phthalic acid, bis-methyl ester D4 |
| CAS | 93951-89-4 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C10 2H4 H6 O4 |
| Mængde | 2.5 mg |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-dimethyl ester, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dimethyl ester (9CI), 1,2-Dimethyl 1,2-benzene-3,4,5,6-d4-dicarboxylate, Phthalic acid-d4 dimethyl ester, Phthalic acid dimethyl ester-d4, Phthalic acid dimethyl ester D4 (phenyl-3,4,5,6-D4), Dimethyl phthalate-3,4,5,6-d4, Phthalic acid, bis-methyl ester D4, Phthalic Acid, Dimethyl Ester D4 (phenyl D4) |
| InChI formel | InChI=1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3/i3D,4D,5D,6D |
| SMIL | [2H]c1c([2H])c([2H])c(C(=O)OC)c(C(=O)OC)c1[2H] |
| IUPAC navn | dimethyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molekylvægt (g/mol) | 198.21 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?