missing translation for 'onlineSavingsMsg'
Få mere at vide

Dimethyl phthalate-3,4,5,6-d4, TRC

Artikelnummer. 30378464
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378464 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378464 Leverandør Toronto Research Chemicals Leverandørnr. TRCD48017825MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Phthalic acid, bis-methyl ester D4
CAS 93951-89-4
Renhedsgrad noter HPLC
Molekylær formel C10 2H4 H6 O4
Mængde 25 mg
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-dimethyl ester, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dimethyl ester (9CI), 1,2-Dimethyl 1,2-benzene-3,4,5,6-d4-dicarboxylate, Phthalic acid-d4 dimethyl ester, Phthalic acid dimethyl ester-d4, Phthalic acid dimethyl ester D4 (phenyl-3,4,5,6-D4), Dimethyl phthalate-3,4,5,6-d4, Phthalic acid, bis-methyl ester D4, Phthalic Acid, Dimethyl Ester D4 (phenyl D4)
InChI formel InChI=1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OC)c(C(=O)OC)c1[2H]
IUPAC navn dimethyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 198.21
Formel vægt 198.083
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.