Få mere at vide
Dimethyl Phosphate-13C2 Sodium Salt, TRC
Artikelnummer. 30135192
Skift visning
Mængde:
0.5 mg
2.5 mg
Pakningsstørrelse:
0.5mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30135192 2.5 mg 2.5mg
30009808 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135192 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4776032.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 157487-95-1
Molekylær formel 13C2H6NaO4P
Molekylvægt (g/mol) 150.02
Synonym Phosphoric Acid Dimethyl Ester-13C2 Sodium Salt, Dimethyl-13C2 Monosodium Phosphate, Dimethyl-13C2 Sodium Phosphate, Monosodium Dimethyl-13C2 Phosphate, Sodium Dimethyl-13C2 Phosphate;
IUPAC navn sodium di(methyl-13C) phosphate
SMIL O=P(O[13CH3])(O[Na])O[13CH3]
Tekniske data
Kemisk navn eller materiale Sodium Dimethyl Phosphate 13C2
Mængde 2.5 mg
InChI formel InChI=1S/C2H7O4P.Na/c1-5-7(3,4)6-2;/h1-2H3,(H,3,4);/q;+1/p-1/i1+1,2+1;
Formel vægt 150