Få mere at vide
Dimethyl Malonate-d2, TRC
Artikelnummer. 30372550
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30372550 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372550 Leverandør Toronto Research Chemicals Leverandørnr. TRCD474592100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dimethyl Malonate-d2
CAS 36647-07-1
Molekylær formel C5H6D2O4
Mængde 100 mg
Synonym 2,2-Dideuteriomalonic Acid Dimethyl Ester
InChI formel InChI=1S/C5H8O4/c1-8-4(6)3-5(7)9-2/h3H2,1-2H3/i3D2
SMIL COC(C([2H])([2H])C(OC)=O)=O
IUPAC navn dimethyl malonate-d2
Molekylvægt (g/mol) 134.13
Formel vægt 134.05