missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl L-tartrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Dimethyl L-tartrate is a chiral auxiliary used for the enantioselective epoxidation of allylic alcohols. It is used for asymmetric α-halogenation of acetals. It is a starting material for the synthesis of chiral building blocks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl L-tartrate is a chiral auxiliary used for the enantioselective epoxidation of allylic alcohols. It is used for asymmetric α-halogenation of acetals. It is a starting material for the synthesis of chiral building blocks.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Dimethyl L-tartrate is a chiral auxiliary used for the enantioselective epoxidation of allylic alcohols. It is used for asymmetric α-halogenation of acetals. It is a starting material for the synthesis of chiral building blocks.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 56°C to 61°C |
Density | 1.3 |
Boiling Point | 158°C (11 mmHg) |
Flash Point | >170°C (338°F) |
Quantity | 25 g |
Beilstein | 1726256 |
Solubility Information | Soluble in water. |
Optical Rotation | +21° (c=1 in Water) |
Formula Weight | 178.14 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'