missing translation for 'onlineSavingsMsg'
Få mere at vide

Dimethyl Hexanedioate-3,3,4,4-d4, CDN

Artikelnummer. 30323529
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323529 0.5 g 0.5g
30318188 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323529 Leverandør CDN Leverandørnr. CDND68130.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Hexanedioic Acid Dimethyl Ester-3,3,4,4-D4
CAS 55724-08-8
Molekylær formel C8 D4 H10 O4
Mængde 0.5 g
Synonym Hexanedioic-3,3,4,4-d4 acid, dimethyl ester (9 CI), 3,3,4,4-Tetradeuterio-hexanedioic acid dimethyl ester, 1,6-Dimethyl hexanedioate-3,3,4,4-d4, Dimethyl Hexanedioate-3,3,4,4-d4, Adipic-3,3,4,4-d4 acid dimethyl ester, Hexanedioic Acid Dimethyl Ester-3,3,4,4-D4
InChI formel InChI=1 S/C8H14O4/c1-11-7(9)5-3-4-6-8(10)12-2/h3-6H2,1-2H3/i3D2,4D2
SMIL [2 H]C([2 H])(CC(=O)OC)C([2 H])([2 H])CC(=O)OC
IUPAC navn dimethyl 3,3,4,4-tetradeuteriohexanedioate
Molekylvægt (g/mol) 178.219
Formel vægt 178.1143 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.