Få mere at vide
Dimethyl Fumarate-d6, TRC
Artikelnummer. 30370032
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30370032 50 mg
30373266 10 mg
30383051 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30370032 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46496650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 66487-95-4
Molekylær formel C6H2D6O4
Molekylvægt (g/mol) 150.16
Synonym (2E)-2-Butenedioic Acid 1,4-Dimethyl Ester-d6, (E)-2-Butenedioic Acid 1,4-Dimethyl Ester-d6, trans-1,2-Ethylenedicarboxylic Acid, Fumaric Acid Dimethyl Ester-d6, Di(methyl-d3) Ester (2E)-2-Butenedioic Acid, Di(methyl-d3) Ester (E)-2-Butenedioic Acid
SMIL [2H]C([2H])([2H])OC(=O)\C=C\C(=O)OC([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale Dimethyl Fumarate-d6
Mængde 50 mg
InChI formel InChI=1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3+/i1D3,2D3
Formel vægt 150.0799