missing translation for 'onlineSavingsMsg'
Få mere at vide

Dimethyl Fumarate-d2, TRC

Artikelnummer. 30376996
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376996 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376996 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46496710MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Dimethyl Fumarate-d2
CAS 23057-98-9
Renhedsgrad noter HPLC
Molekylær formel C6 D2 H6 O4
Mængde 10 mg
Synonym 2-Butenedioic-2,3-d2 acid, 1,4-dimethyl ester, (2E)- (ACI), 1,4-Dimethyl (2E)-2-butenedioate-2,3-d2 (ACI), 2-Butenedioic-2,3-d2 acid, dimethyl ester, (2E)- (9CI), 2-Butenedioic-2,3-d2 acid, dimethyl ester, (E)- (ZCI), Fumaric-d2 acid, dimethyl ester (8CI), Dimethyl fumarate-d2, Dimethyl Fumarate-2,3-D2, (2E)-2-Butenedioic-2,3-d2 acid dimethyl ester
InChI formel InChI=1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3+/i3D,4D
SMIL [2H]\C(=C(\[2H])/C(=O)OC)\C(=O)OC
IUPAC navn dimethyl (E)-2,3-dideuteriobut-2-enedioate
Molekylvægt (g/mol) 146.138
Formel vægt 146.055
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.