Learn More
Dimethyl Disulfide, Sigma-Aldrich™
Product Code. 15884561 Shop All Merck Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884561 1 L 1 L
15874561 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15884561 Supplier Sigma Aldrich Supplier No. 8207911000
 Restricted Item
Forventet forsendelse: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 624-92-0
Specifications
Melting Point -85°C
Density 1.06 g/cm3 at 20°C
MDL Number MFCD00008561
Quantity 1 L
Synonym Dimethyl disulfide, Methyl disulfide, Dimethyl disulfide
Molecular Weight (g/mol) 94.2
Grade Research