Få mere at vide
Dimethyl-d6 Trisulfide, TRC
Artikelnummer. 30371932
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30371932 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371932 Leverandør Toronto Research Chemicals Leverandørnr. TRCD479672100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dimethyl-d6 Trisulfide
CAS 58069-93-5
Molekylær formel C2D6S3
Mængde 100 mg
Synonym Di(methyl-d3) Trisulfide
InChI formel InChI=1S/C2H6S3/c1-3-5-4-2/h1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])SSSC([2H])([2H])[2H]
Molekylvægt (g/mol) 132.3