Få mere at vide
Dimethyl (4S)-4-Methyl-(2-oxooctyl)phosphonate-d3, TRC
Artikelnummer. 30378439
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30378439 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378439 Leverandør Toronto Research Chemicals Leverandørnr. TRCD474752250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dimethyl (4S)-4-Methyl-(2-oxooctyl)phosphonate-d3
CAS 1329642-51-4
Molekylær formel C11H20D3O4P
Mængde 250 mg
Synonym (S)-(4-Methyl-2-oxooctyl)phosphonic Acid Dimethyl Ester-d3, [(4S)-4-Methyl-2-oxooctyl]phosphonic Acid Dimethyl Ester-d3
InChI formel InChI=1S/C11H23O4P/c1-5-6-7-10(2)8-11(12)9-16(13,14-3)15-4/h10H,5-9H2,1-4H3/t10-/m0/s1/i2D3
SMIL [2H]C([2H])([2H])[C@@H](CCCC)CC(=O)CP(=O)(OC)OC
Molekylvægt (g/mol) 253.29
Formel vægt 253.1522