Få mere at vide
Dimethyl (3-phenoxy-2-oxoproyl)phosphonate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dimethyl (3-phenoxy-2-oxoproyl)phosphonate |
| CAS | 40665-68-7 |
| Molekylær formel | C11H15O5P |
| Mængde | 50 g |
| Synonym | P-(2-oxo-3-Phenoxypropyl)phosphonic Acid Dimethyl Ester, (2-oxo-3-Phenoxypropyl)phosphonic Acid Dimethyl Ester, (2-Oxo-3-phenoxypropyl)phosphonic Acid Dimethyl Ester, Dimethyl (2-oxo-3-phenoxypropyl)phosphonate, Dimethyl (3-Phenoxyacetonyl)phosphonate, Dimethyl 3-{Phenoxy-2-oxopropylphosphonate |
| InChI formel | InChI=1S/C11H15O5P/c1-14-17(13,15-2)9-10(12)8-16-11-6-4-3-5-7-11/h3-7H,8-9H2,1-2H3 |
| SMIL | COP(=O)(CC(=O)COc1ccccc1)OC |
| Molekylvægt (g/mol) | 258.21 |