Få mere at vide
Dimethyl (2S)-2-[[4-[[(2,4-Diaminopteridin-6-yl)methyl]methylamino]benzoyl]amino]pentanedioate (Methotrexate Dimethyl Ester), Mikromol™
Beskrivelse
Dimethyl (2S)-2-[[4-[[(2,4-Diaminopteridin-6-yl)methyl]methylamino]benzoyl]amino]pentanedioate (Methotrexate Dimethyl Ester)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methotrexate Dimethyl Ester |
| CAS | 34378-65-9 |
| Analyt- eller komponentnavne | Methotrexate Dimethyl Ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H26 N8 O5 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, 1,5-dimethyl ester, Glutamic acid, N-[p-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, dimethyl ester, L-(+)- (8CI), L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, dimethyl ester (9CI), Dimethyl methotrexate, Methotrexate dimethyl ester, Methotrexate Imp. J (EP), Methotrexate Impurity J |
| InChI formel | InChI=1S/C22H26N8O5/c1-30(11-13-10-25-19-17(26-13)18(23)28-22(24)29-19)14-6-4-12(5-7-14)20(32)27-15(21(33)35-3)8-9-16(31)34-2/h4-7,10,15H,8-9,11H2,1-3H3,(H,27,32)(H4,23,24,25,28,29)/t15-/m0/s1 |
| SMIL | COC(=O)CC[C@H](NC(=O)c1ccc(cc1)N(C)Cc2cnc3nc(N)nc(N)c3n2)C(=O)OC |
| IUPAC navn | dimethyl (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioate |
| Vis mere |