Få mere at vide
Dimethyl (2-Oxononyl)phosphonate-d15, TRC
Artikelnummer. 30371425
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30371425 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371425 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47645725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dimethyl (2-Oxononyl)phosphonate-d15
CAS 1215800-33-1
Molekylær formel C11H8D15O4P
Mængde 25 mg
Synonym (2-Oxononyl)phosphonic Acid Dimethyl Ester-d15
InChI formel InChI=1S/C11H23O4P/c1-4-5-6-7-8-9-11(12)10-16(13,14-2)15-3/h4-10H2,1-3H3/i1D3,4D2,5D2,6D2,7D2,8D2,9D2
SMIL O=P(CC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)(OC)OC
IUPAC navn dimethyl (2-oxononyl-3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-d15)phosphonate
Molekylvægt (g/mol) 265.36
Formel vægt 265.23