Få mere at vide
Dimethyl 1,10-decanedicarboxylate, 98%, Thermo Scientific™
Artikelnummer. 11362696
Skift visning
Mængde:
10 g
50 g
Pakningsstørrelse:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11362696 50 g 50g
11352696 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11362696 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L08757.18
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1731-79-9
Molekylær formel C14H26O4
Molekylvægt (g/mol) 258.358
MDL nummer MFCD00043632
InChI nøgle IZMOTZDBVPMOFE-UHFFFAOYSA-N
Synonym dimethyl 1,10-decanedicarboxylate, dodecanedioic acid dimethyl ester, dodecanedioic acid, dimethyl ester, dimethyl 1,12-dodecanedioate, 1,12-dimethyl dodecanedioate, dodecanedioic acid dimethyl, dimethyl dodecane-1,12-dioate, dimethyldodecanedioate, dodecanedioic acid, 1,12-dimethyl ester, acmc-209e5w
PubChem CID 74415
IUPAC navn dimethyl dodecanedioate
SMIL COC(=O)CCCCCCCCCCC(=O)OC
Tekniske data
Kemisk navn eller materiale Dimethyl 1,10-decanedicarboxylate
Smeltepunkt 27°C to 32°C
Kogepunkt 187°C to 188°C (14 mmHg)
Lineær formel CH3O2C(CH2)10CO2CH3
Mængde 50 g
Beilstein 1790424
Formel vægt 258.36
Procent renhed 98%