Få mere at vide
Dimethyl-1,1,1-d3-amine HCl, CDN
Artikelnummer. 30311451
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311451 0.25 g
30325920 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311451 Leverandør CDN Leverandørnr. CDND25440.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dimethylamine methyl-d3 hydrochloride
CAS 120033-84-3
Molekylær formel C2 2H3 H4 N . Cl H
Mængde 0.25 g
InChI formel InChI=1 S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1 H/i1D3;
SMIL Cl.[2 H]C([2 H])([2 H])NC
IUPAC navn 1,1,1-trideuterio-N-methylmethanamine;hydrochloride
Molekylvægt (g/mol) 84.56
Formel vægt 84.0534 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity