Få mere at vide
Dimethrin, TRC
Artikelnummer. 30009657
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009657 100 mg
30006387 10 mg
30008984 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009657 Leverandør Toronto Research Chemicals Leverandørnr. TRCD303035100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70-38-2
Molekylær formel C19 H26 O2
Molekylvægt (g/mol) 286.41
Synonym Chrysanthemummonocarboxylic acid, 2,4-dimethylbenzyl ester (6CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, (2,4-dimethylphenyl)methyl ester (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)-, 2,4-dimethylbenzyl ester (7CI,8CI), 2,4-Dimethylbenzyl 2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylate, 2,4-Dimethylbenzyl chrysanthemumate, Chrysanthemumic acid 2,4-dimethylbenzyl ester, Dimethrin, Dimetrin, ENT 21,170, NSC 15731
IUPAC navn (2,4-dimethylphenyl)methyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
SMIL CC(=CC1C(C(=O)OCc2ccc(C)cc2C)C1(C)C)C
Tekniske data
Kemisk navn eller materiale Dimethrin
Mængde 100 mg
InChI formel InChI=1S/C19H26O2/c1-12(2)9-16-17(19(16,5)6)18(20)21-11-15-8-7-13(3)10-14(15)4/h7-10,16-17H,11H2,1-6H3
Formel vægt 286.1933