Få mere at vide
Dimethoxanate Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dimethoxanate Hydrochloride |
| CAS | 518-63-8 |
| Analyt- eller komponentnavne | Dimethoxanate Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H22 N2 O3 S . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 10H-Phenothiazine-10-carboxylic acid, 2-[2-(dimethylamino)ethoxy]ethyl ester, hydrochloride (1:1), 10H-Phenothiazine-10-carboxylic acid, 2-[2-(dimethylamino)ethoxy]ethyl ester, monohydrochloride (9CI), Phenothiazine-10-carboxylic acid 2-[2-(dimethylamino)ethoxy]ethyl ester hydrochloride (6CI), Phenothiazine-10-carboxylic acid, 2-[2-(dimethylamino)ethoxy]ethyl ester, monohydrochloride (7CI,8CI), Ethanol, 2-[2-(dimethylamino)ethoxy]-, phenothiazine-10-carboxylate (ester), hydrochloride (8CI), 2-Dimethylaminoethoxyethyl 10-phenothazinecarboxylate hydrochloride, Cothera, Dimethoxanate hydrochloride, Dimethoxanate monohydrochloride, Dimetoxanate hydrochloride, β-Dimethylaminoethoxyethyl phenothiazine-10-carboxylate hydrochloride |
| InChI formel | InChI=1S/C19H22N2O3S.ClH/c1-20(2)11-12-23-13-14-24-19(22)21-15-7-3-5-9-17(15)25-18-10-6-4-8-16(18)21;/h3-10H,11-14H2,1-2H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CN(C)CCOCCOC(=O)N1c2ccccc2Sc3ccccc13 |
| Vis mere |