Få mere at vide
Dimethachlor CGA 369873, TRC
Artikelnummer. 30138076
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138076 250 mg
30004677 100 mg
30010260 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138076 Leverandør Toronto Research Chemicals Leverandørnr. TRCD465480250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2387071-47-6
Molekylær formel C10 H12 N O4 S . Na
Molekylvægt (g/mol) 265.26
Synonym Dimethachlor Metabolite CGA 369873 Sodium
IUPAC navn sodium;2-(2,6-dimethylanilino)-2-oxoethanesulfonate
SMIL [Na+].Cc1cccc(C)c1NC(=O)CS(=O)(=O)[O-]
Tekniske data
Kemisk navn eller materiale Dimethachlor Metabolite CGA 369873 Sodium
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C10H13NO4S.Na/c1-7-4-3-5-8(2)10(7)11-9(12)6-16(13,14)15;/h3-5H,6H2,1-2H3,(H,11,12)(H,13,14,15);/q;+1/p-1
Formel vægt 265.0385
Procent renhed >95%