missing translation for 'onlineSavingsMsg'
Få mere at vide

Dimenhydrinate EP Impurity F (Diphenhydramine EP Impurity A) as Hydrochloride, Mikromol™

Artikelnummer. 30052513
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052513 25 mg
17962416 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30052513 Leverandør Mikromol Leverandørnr. MM0490.030025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dimenhydrinate EP Impurity F (Diphenhydramine EP Impurity A) as Hydrochloride
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Nordiphenhydramine Hydrochloride
CAS 53499-40-4
Analyt- eller komponentnavne Nordiphenhydramine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C16 H19 N O . Cl H
Certificeringer/compliance ISO 17025
Synonym 2-(Diphenylmethoxy)-N-methylethanamine Hydrochloride (Nordiphenhydramine Hydrochloride), Dimenhydrinate Imp. F (EP), Diphenhydramine Hydrochloride Imp. A (EP), Diphenhydramine Imp. A (EP), Diphenhydramine USP Related Compound A, Diphenhydramine USP RC A, 2-(Diphenylmethoxy)-N-methylethanamine Hydrochloride, Nordiphenhydramine Hydrochloride, Dimenhydrinate Imp. F (EP) as Hydrochloride, Diphenhydramine Imp. A Hydrochloride, Diphenhydramine Hydrochloride Imp. A (EP) as Hydrochloride, Diphenhydramine Related Compound A as Hydrochloride, Dimenhydrinate Impurity F as Hydrochloride, Diphenhydramine Hydrochloride Impurity A as Hydrochloride, Diphenhydramine Impurity A as Hydrochloride
InChI formel InChI=1S/C16H19NO.ClH/c1-17-12-13-18-16(14-8-4-2-5-9-14)15-10-6-3-7-11-15;/h2-11,16-17H,12-13H2,1H3;1H
SMIL Cl.CNCCOC(c1ccccc1)c2ccccc2
IUPAC navn 2-benzhydryloxy-N-methylethanamine;hydrochloride
Molekylvægt (g/mol) 277.79
Mængde 25 mg
Formel vægt 277.1233 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.