Få mere at vide
DIM-C-pPhOCH3, Tocris Bioscience™
Nur77 agonist
Brand: Tocris Bioscience 6376/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
DIM-C-pPhOCH3 is a Nur77 agonist. Induces apoptosis in cancer cell lines in vitro. Enhances long-term potentiation in hippocampal slices and long-term contextual fear memory in an in vivo mouse model.Tekniske data
| 1,1-Bis(3'-indolyl)-1-(p-methoxyphenyl)methane | |
| 33985-68-1 | |
| >98% | |
| 5 mg |