missing translation for 'onlineSavingsMsg'
Få mere at vide

DIM-C-pPhOCH3

Artikelnummer. 18730979
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18730979 5 mg
18729278 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18730979 Leverandør Tocris Bioscience Leverandørnr. 6376/5

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Nur77 agonist

DIM-C-pPhOCH3 is a Nur77 agonist. Induces apoptosis in cancer cell lines in vitro. Enhances long-term potentiation in hippocampal slices and long-term contextual fear memory in an in vivo mouse model.
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1,1-Bis(3'-indolyl)-1-(p-methoxyphenyl)methane
CAS 33985-68-1
Analyseprocentområde >98%
Mængde 5 mg
Molekylær formel C24H20N2O
Formel vægt Observed MW: 352.43
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.