Få mere at vide
DIM-C-pPhOCH3
Artikelnummer. 18729278
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18729278 25 mg
18730979 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18729278 Leverandør Tocris Bioscience Leverandørnr. 6376/25
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Nur77 agonist
DIM-C-pPhOCH3 is a Nur77 agonist. Induces apoptosis in cancer cell lines in vitro. Enhances long-term potentiation in hippocampal slices and long-term contextual fear memory in an in vivo mouse model.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1-Bis(3'-indolyl)-1-(p-methoxyphenyl)methane
CAS 33985-68-1
Analyseprocentområde >98%
Mængde 25 mg
Molekylær formel C24H20N2O
Formel vægt Observed MW: 352.43