Få mere at vide
Diltiazem-d3 Hydrochloride, TRC
Artikelnummer. 30370990
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30370990 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370990 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46062250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diltiazem-d3 Hydrochloride
CAS 1217623-80-7
Renhedsgrad noter HPLC
Molekylær formel C22H24D3ClN2O4S
Mængde 50 mg
Synonym (2S,cis)-3-(Acetyloxy)-5-[(2-dimethylamino-d3)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, CRD-401-d3, Adizem-d3, Altiazem-d3, Dilzem-d3, Masdil-d3, Zilden-d3
InChI formel InChI=1S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21H,13-14H2,1-4H3;1H/t20-,21+;/m1./s1/i2D3;
SMIL O=C(N(CCN(C([2H])([2H])[2H])C)C(C=CC=C1)=C1S2)[C@H](OC(C)=O)[C@@H]2C3=CC=C(OC)C=C3.Cl
Molekylvægt (g/mol) 454
Procent renhed >95